Dropping bottle with head stopper
Supplier: MARIENFELD
3610105
MARI3610105EA 0 SGD
MARI3610105
Dropping bottle with head stopper
Bottles Dropping Bottles
Glass, amber.
  • Good chemical and physical properties
  • For use in applications that do not involve high thermal stresses
  • Amber coloured to protect light sensitive substanzes
Dropping bottle with flat head stopper and round shoulder.