Preparation table for slides
Supplier: MARIENFELD
6000007
MARI6000007EA 0 SGD
MARI6000007
Preparation table for slides
Slide Holders
Preparation table for 20 microscopy slides. Thanks to its recessed grips the slides can be easily taken out.