Laboratory marker
Supplier: MARIENFELD
6130603
MARI6130603EA 7.7 SGD
MARI6130603
Laboratory marker
Pens Lab Markers
Permanent marker for indelibly labelling microscope slides and embedding cassettes with a high contrast black ink. Ultra-fine tip. These laboratory markers do not dry out and have a long lifetime.
  • Water resistant, odour-free and non toxic
  • Resistant against most laboratory chemicals
  • No pretreatment of surfaces necessary