Digital stopwatch, STRATOS 2
Supplier: MARIENFELD
6390110
MARI6390110EA 0 SGD
MARI6390110
Digital stopwatch, STRATOS 2
Timers Digital Stopwatches
Electronic timer with two buttons have the functions of start/stop/reset, addition/split/dual measuring, with 7-digits LCD display (height of numbers: ~6,5 mm).
  • Display range: 9 H, 59 min and 59,99 s, ¹/₁₀₀ s
  • With plastic housing and lanyard
  • With battery (button cell SR 54)