Racks and lids for storage 2D barcoded tubes, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
4890
MATR4890EA 0 SGD
MATR4890
Racks and lids for storage 2D barcoded tubes, Matrix™
Racks Cryogenic Vial Racks
Compatible with automated systems and rack features locking lid to secure samples.
  • Made of plastic
  • Superior storage rack design
  • Latch racks to save precious space
  • Color: white
The latch rack design enables manual multichannel pipette access to 2D tubes and eliminates contamination risk with lid design that does not contact the benchtop. The lid can be lifted robotically for 2D tube access with automated liquid handling systems and high throughput applications.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement