Deep well storage plates, 96-well, 0,8 ml
Supplier: THERMO MATRIX TECHNOLOGIES
AB-0765 AB-0859
MATRAB-0765EA 0 SGD
MATRAB-0765 MATRAB-0859
Deep well storage plates, 96-well, 0,8 ml
Microplates Deep Well Microplates
PP. 96-well plates for sample storage.
  • 0,8 ml working volume when sealed with adhesive or heat seals, 0,7 ml when sealed with storage plate cap strips or 0,55 ml when sealed with a 96-cap sealing mat
  • Conical well bottom for ease of sample retrieval
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification IRECS; ISO 14001 (Certificate Registration No. EMS 752603)