CO₂ incubators, ICO series

Supplier: MEMMERT
ICO50 ICO150
MEMMICO50EA 0 SGD
MEMMICO50 MEMMICO150
CO₂ incubators, ICO series
Incubators CO₂ Incubators
The ICO series of CO₂ incubators has seamlessly welded, corrosion-resistant stainless steel chambers with rounded edges and no further installations, ensuring easy and thorough cleaning. Models have TwinDISPLAY with a USB port for uploading programs, reading out logs or protecting appliances via the user ID function, and an Ethernet interface as well as a data logger with a 10 year storage capacity. All parameters can be set directly in the ControlCOCKPIT or the AtmoCONTROL software, which uses drag and drop symbols to input values. Battery-buffered ControlCOCKPIT (optional): OperatingMore Product Information
Order Now

SPECIFICATIONS

Model ICO50 ICO150
Environmentally Preferable
Temperature fluctuation (time) ±0,1 °C @ 37 °C ±0,1 °C @ 37 °C
Temperature variation (spatial) ±0,3 °C @ 37 °C ±0,3 °C @ 37 °C
Recovery time after 30 second door opening at 37 °C 3 min 5 min
Capacity 56 L 156 L
Usable volume 19,67 L 69,89 L
Convection type Forced convection Forced convection
CO₂ recovery time after 30 second door opening at 5 Vol.% CO₂ - 8 min
Shelves supplied (max.) 0 (5) 0 (10)
Max. load per shelf 15 kg 15 kg
Max. chamber workload 75 kg 120 kg
Int. W×D×H 400×330×425 mm 560×400×700 mm
Ext. W×D×H 559×521×791 mm 719×591×1066 mm
Energy consumption 60 Wh/h at 37 °C 80 Wh/h at 37 °C
Nominal power consumption 1000 W 2000 W
Sound level 50 dB(A) 50 dB(A)
Temperature range Ambient +5…50 °C Ambient +5…50 °C
Weight 55 kg 90 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR