CO₂ incubators, ICOmed series
Supplier: MEMMERT
ICO105MED ICO150MED ICO240MED ICO50MED
MEMMICO105MEDEA 0 SGD
MEMMICO105MED MEMMICO150MED MEMMICO240MED MEMMICO50MED
CO₂ incubators, ICOmed series
Incubators CO₂ Incubators
The ICOmed series of CO₂ incubators has seamlessly welded, corrosion-resistant stainless steel chambers with rounded edges and no further installations, ensuring easy and thorough cleaning. Models have TwinDISPLAY with a USB port for uploading programs, reading out logs or protecting appliances via the user ID function, and an Ethernet interface as well as a data logger with a ten year storage capacity. All parameters can be set directly in the ControlCOCKPIT or the AtmoCONTROL software, which uses drag and drop symbols to input values. Battery-buffered ControlCOCKPIT (optional): OperatingMore Product Information