Heating and drying ovens, Universal UF/UFplus (forced convection)
Supplier: MEMMERT
UF55 UF55PLUS UF75 UF750 UF750PLUS UF75PLUS UF110 UF110PLUS UF160 UF160PLUS UF260 UF260PLUS UF30 UF30PLUS UF450 UF450PLUS UF1060 UF1060PLUS UF1060MPLUS UF110M UF110MPLUS UF55M UF160M UF55MPLUS UF160MPLUS UF75M UF260M UF75MPLUS UF260MPLUS UF750M UF30M UF750MPLUS UF30MPLUS UF450M UF450MPLUS UF1060M
MEMMUF55EA 4810 SGD
MEMMUF55 MEMMUF55PLUS MEMMUF75 MEMMUF750 MEMMUF750PLUS MEMMUF75PLUS MEMMUF110 MEMMUF110PLUS MEMMUF160 MEMMUF160PLUS MEMMUF260 MEMMUF260PLUS MEMMUF30 MEMMUF30PLUS MEMMUF450 MEMMUF450PLUS MEMMUF1060 MEMMUF1060PLUS MEMMUF1060MPLUS MEMMUF110M MEMMUF110MPLUS MEMMUF55M MEMMUF160M MEMMUF55MPLUS MEMMUF160MPLUS MEMMUF75M MEMMUF260M MEMMUF75MPLUS MEMMUF260MPLUS MEMMUF750M MEMMUF30M MEMMUF750MPLUS MEMMUF30MPLUS MEMMUF450M MEMMUF450MPLUS MEMMUF1060M
Heating and drying ovens, Universal UF/UFplus (forced convection)
Ovens Standard Ovens
The range includes forced convection models with a temperature range up to 300 °C. There is a choice of two different displays, SingleDISPLAY or TwinDISPLAY depending on the application.
  • SingleDISPLAY models, ideal for standard applications:
  • ControlCOCKPIT parameters: Temperature (Celsius or Fahrenheit), fan speed, exhaust air flap position, program time, time zones and daylight saving time
  • One PT100 temperature sensor in a four wire circuit and Ethernet interface for reading out the protocol log
  • Double over-temperature protection including visual
More Product Information
Order Now
SPECIFICATIONS
Model UF55 UF55plus UF75 UF750 UF750plus UF75plus UF110 UF110plus UF160 UF160plus UF260 UF260plus UF30 UF30plus UF450 UF450plus UF1060 UF1060plus UF1060mplus UF110m UF110mplus UF55m UF160m UF55mplus UF160mplus UF75m UF260m UF75mplus UF260mplus UF750m UF30m UF750mplus UF30mplus UF450m UF450mplus UF1060m
Environmentally Preferable
Temperature fluctuation (time) @ 150 °C: ≤±0,5 °C @ 150 °C: ≤±0,5 °C @ 150 °C: ≤±0,5 °C @ 150 °C: ≤±0,6 °C @ 150 °C: ≤±0,6 °C @ 150 °C: ≤±0,5 °C @ 150 °C: ≤±0,5 °C @ 150 °C: ≤±0,5 °C @ 150 °C: ≤±0,6 °C @ 150 °C: ≤±0,6 °C @ 150 °C: ≤±0,5 °C @ 150 °C: ≤±0,5 °C @ 150 °C: ≤±0,5 °C @ 150 °C: ≤±0,5 °C @ 150 °C: ≤±0,6 °C @ 150 °C: ≤±0,6 °C @ 150 °C: ≤±0,8 °C @ 150 °C: ≤±0,8 °C @ 150 °C: ≤±0,8 °C @ 150 °C: ≤±0,5 °C @ 150 °C: ≤±0,5 °C @ 150 °C: ≤±0,5 °C @ 150 °C: ≤±0,6 °C @ 150 °C: ≤±0,5 °C @ 150 °C: ≤±0,6 °C @ 150 °C: ≤±0,5 °C @ 150 °C: ≤±0,5 °C @ 150 °C: ≤±0,5 °C @ 150 °C: ≤±0,5 °C @ 150 °C: ≤±0,6 °C @ 150 °C: ≤±0,5 °C @ 150 °C: ≤±0,6 °C @ 150 °C: ≤±0,5 °C @ 150 °C: ≤±0,6 °C @ 150 °C: ≤±0,6 °C @ 150 °C: ≤±0,8 °C
Temperature range ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C ambient +10…300 °C
Temperature variation (spatial) ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C ≤ ±2 °C @ 150 °C
Capacity 53 L 53 L 74 L 749 L 749 L 74 L 108 L 108 L 161 L 161 L 256 L 256 L 32 L 32 L 449 L 449 L 1060 L 1060 L 1060 L 108 L 108 L 53 L 161 L 53 L 161 L 74 L 256 L 74 L 256 L 749 L 32 L 749 L 32 L 449 L 449 L 1060 L
Convection type Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection Forced convection
Max. load per shelf 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg 30 kg
Power consumption 2000 W 2000 W 2500 W 7000 W 7000 W 2500 W 2800 W 2800 W 3200 W 3200 W 3400 W 3400 W 1600 W 1600 W 5800 W 5800 W 7000 W 7000 W 7000 W 2800 W 2800 W 2000 W 3200 W 2000 W 3200 W 2500 W 3400 W 2500 W 3400 W 7000 W 1600 W 7000 W 1600 W 5800 W 5800 W 7000 W