Heating and drying ovens, Universal UN/UNplus (natural convection)
Supplier: MEMMERT
UN75 UN750 UN750PLUS UN75PLUS UN110 UN110PLUS UN160 UN160PLUS UN30 UN260 UN260PLUS UN30PLUS UN450 UN450PLUS UN55 UN55PLUS UN260M UN260MPLUS UN30M UN30MPLUS UN450M UN450MPLUS UN750M UN110M UN750MPLUS UN110MPLUS UN160M UN55M UN160MPLUS UN55MPLUS UN75M UN75MPLUS
MEMMUN75EA 4540 SGD
MEMMUN75 MEMMUN750 MEMMUN750PLUS MEMMUN75PLUS MEMMUN110 MEMMUN110PLUS MEMMUN160 MEMMUN160PLUS MEMMUN30 MEMMUN260 MEMMUN260PLUS MEMMUN30PLUS MEMMUN450 MEMMUN450PLUS MEMMUN55 MEMMUN55PLUS MEMMUN260M MEMMUN260MPLUS MEMMUN30M MEMMUN30MPLUS MEMMUN450M MEMMUN450MPLUS MEMMUN750M MEMMUN110M MEMMUN750MPLUS MEMMUN110MPLUS MEMMUN160M MEMMUN55M MEMMUN160MPLUS MEMMUN55MPLUS MEMMUN75M MEMMUN75MPLUS
Heating and drying ovens, Universal UN/UNplus (natural convection)
Ovens Standard Ovens
The range includes natural convection models with a temperature range up to +300 °C. There is a choice of two different displays, SingleDISPLAY or TwinDISPLAY depending on the application.
  • SingleDISPLAY models, ideal for standard applications:
  • ControlCOCKPIT parameters: Temperature (Celsius or Fahrenheit), fan speed, exhaust air flap position, program time, time zones and daylight saving time
  • One PT100 temperature sensor in a four wire circuit and Ethernet interface for reading out the protocol log
  • Double over-temperature protection including visual
More Product Information