Microscope slides, SuperFrost®, Menzel Gläser
Supplier: Epredia
AG00008532E01 AG00008632E01 AA00008132E01 AG00008432E012 AG00008032E012 AG00008132E012 AG00008232E20 AA00008032E011
MENZAG00008532E01EA 13.1 SGD
MENZAG00008532E01 MENZAG00008632E01 MENZAA00008132E01 MENZAG00008432E012 MENZAG00008032E012 MENZAG00008132E012 MENZAG00008232E20 MENZAA00008032E011
Microscope slides, SuperFrost®, Menzel Gläser
Slides Microscope Slides
These slides are made of extra-white glass, washed clean and polished.
  • Labelling area resistant to all common solvents
  • Six colours for identification and colour coding
  • Thick labelling area to prevent slides from sticking together or getting scratched
Suitable for in vitro diagnostic (IVD) applications.
Complies with ISO 8037/I
Certifications: ISO 8037/I.
Packaging: In cardboard boxes of 50.
Order Now
SPECIFICATIONS
Environmentally Preferable