Adhesion slides, Menzel Gläser, Polysine®
Supplier: Epredia
J2800AMNZ
MENZJ2800AMNZEA 54.2 SGD
MENZJ2800AMNZ
Adhesion slides, Menzel Gläser, Polysine®
Slides Adhesive Slides
These slides, made using a technology that electrostatically and chemically attracts fresh cell preparations, formalin-fixed Bouin's or alcohol-fixed paraffin-embedded tissues.
  • 90° ground edges with frosted ends for labelling
  • Adhesion superior to glue-, protein- or silane-treated slides
  • Thick tissue sections of 2 to 5 µm
Unaffected by enzyme pre-digestion or heating, tissue loss is minimised in prolonged in-situ DNA hybridisation techniques and in immunocytochemical methods.
Order Now
SPECIFICATIONS
Environmentally Preferable