Cavity slides
Supplier: Epredia
X1XER315L X2XER208B X1XER301B X1XER302W X2XER203B X1XER308B X1XER305B X1XER303B X1XER311B
MENZX1XER315LEA 0 SGD
MENZX1XER315L MENZX2XER208B MENZX1XER301B MENZX1XER302W MENZX2XER203B MENZX1XER308B MENZX1XER305B MENZX1XER303B MENZX1XER311B
Cavity slides
Slides Cavity Slides
These autoclavable slides are specially designed for immunofluorescence.
  • Adhesive surface and hydrophobic mask
  • Hydrophobic epoxy- or PTFE-coating
  • (*) with rings or wells
Developped for clinical diagnosis applications.
Ordering information: Available with rings or wells (*).