Masterflex® B/T® PerfectPosition® Pump Tubing, Tygon® E-Food, Avantor®
Supplier: Avantor Fluid Handling
MFLA06418-87EA 0 SGD
MFLA06418-87 MFLA06418-91
Masterflex® B/T® PerfectPosition® Pump Tubing, Tygon® E-Food, Avantor®
Tubing Peristaltic Pump Tubing
Ensure optimum performance from your Masterflex® pump.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for
More Product Information