Scienceware Flow Control Hosecock Clamps, Plastic
Supplier: Avantor Fluid Handling
MFLA06833-10EA 70.4 SGD
MFLA06833-10
Scienceware Flow Control Hosecock Clamps, Plastic
Clamps Tubing and Sanitary Clamps
Deliver accurate regulation or interruption of fluid flow.
  • Constructed of Delrin acetal
DELRIN(R) resists weak acids, bases, and organic solvents. Controls flow in flexible tubing up to 1/2" (13 mm) OD. Replaces stopcocks in many applications. Install without disconnecting tubing. Autoclavable. Bel-Art.