Micropump Compact Low-Flow Gear Pump Heads - C Mount Design
Supplier: Avantor Fluid Handling
07002-33
MFLA07002-12EA 0 SGD
MFLA07002-12 MFLA07002-20 MFLA07002-32 MFLA07002-35 MFLA74012-11 MFLB07002-33
Micropump Compact Low-Flow Gear Pump Heads - C Mount Design
Pumps Pump Heads Gear Pump Heads
Offering remote C-mount choices for your low-flow applications.
  • Use for applications requiring high initial differential pressure.
  • Suction-shoe design ensures complete fluid transfer for increased volumetric accuracy
  • Pump efficiency increases as discharge pressure increases
The compact pump head requires a drive for proper operation. Order drives separately.
Optional Service kit includes: gears, suction shoe, shaft seals, and springs. Order separately.
Contact our Application Specialists for assistance.