Masterflex® L/S® and I/P® Pump Head Replacement Parts, Avantor®
Supplier: Avantor Fluid Handling
MFLA07013-80EA 309 SGD
MFLA07013-80 MFLA07013-95 MFLA07014-81 MFLA07015-75 MFLA07015-80 MFLA07015-81 MFLA07015-90 MFLA07015-91 MFLA07016-81 MFLA07017-81 MFLA07017-91 MFLA07018-81 MFLA07018-91 MFLA07019-74 MFLA07019-75 MFLA07019-77 MFLA07019-78 MFLA07019-79 MFLA07019-80 MFLA07019-82 MFLA07019-83 MFLA07019-85 MFLA07019-92 MFLA07024-81 MFLA07024-91 MFLA07036-01 MFLA07036-02 MFLA07514-08 MFLA07514-18 MFLA07516-07 MFLA07516-08 MFLA07516-17 MFLA07516-18 MFLA07518-68 MFLA07518-69 MFLA77602-08 MFLA77602-18 MFLA07013-76 MFLA07013-81 MFLA07013-91 MFLA07014-91 MFLA07016-91 MFLA07019-73 MFLA07035-91 MFLA07514-17 MFLA07514-19
Masterflex® L/S® and I/P® Pump Head Replacement Parts, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Get the most our of your Masterflex® Pump Head with these replacement parts.
  • Choose replacement parts suitable for your L/S® and I/P® pump head
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.