Masterflex® L/S® Multichannel Cartridge Pump Head with 4-Rollers, Avantor®
Supplier: Avantor Fluid Handling
MFLA07519-06EA 1240 SGD
MFLA07519-06
Masterflex® L/S® Multichannel Cartridge Pump Head with 4-Rollers, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Synchronous, multichannel flow from up to 8 channels.
  • Cartridges snap in and out for fast tubing changes—no need to remove pump head from the drive
  • Load pump heads with 1 to 8 cartridges
  • Mix large and small cartridges—one large cartridge occupies the same space as two small ones
  • Flow rate 0.047 to 1400 ml/min with microbore or L/S tubing—flow rate depends on drive rpm, cartridge, and tubing size
  • Accepts 1 to 8 small or 1 to 4 large cartridges
  • Require drives with a minimum of 1⁄10 hp and a maximum of 600 rpm
  • Constructed with stainless steel rotor and
More Product Information