Masterflex® L/S® Multichannel Cartridge Pump Head with Reduced Pulsation, 6-Channel, Avantor®
Supplier: Avantor Fluid Handling
MFLA07519-15EA 2680 SGD
MFLA07519-15
Masterflex® L/S® Multichannel Cartridge Pump Head with Reduced Pulsation, 6-Channel, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Synchronous, low-pulsation flow from multiple channels.
  • Cartridges snap in and out for fast tubing changes—no need to remove pump head from the drive
  • Improved retainer design holds tubing securely in place
  • Flow rate 0.00017 to 170 ml/min with L/S tubing—flow rate depends on drive rpm, cartridge, and tubing size
  • Accepts up to 6 large cartridges
  • Require drives with a minimum of 1⁄10 hp and a maximum of 600 rpm
  • Constructed with 6 Rulon® rollers, stainless steel rotor, and PSF ends
Each pump head system is composed of a heavy-duty pump head, cartridges,More Product Information