Masterflex® Ismatec® 4-Channel Cartridge Pump Head with Stepper Motor, Panel Mount
Supplier: Avantor Fluid Handling
MFLA07519-90EA 0 SGD
MFLA07519-90
Masterflex® Ismatec® 4-Channel Cartridge Pump Head with Stepper Motor, Panel Mount
Pumps Custom Engineered Pumps
Versatile multi-channel panel mount peristaltic pumps provide synchronous and repeatable flow results across four channels.
  • 12-roller design minimises pulsation
  • Click-N-Go cassettes feature spring loaded occlusion for quick tubing changeovers
  • Compact design for efficient use of space
  • Stepper motor allows for precision repeat and rapid dispensing applications
Masterflex panel mount pump systems feature the flexibility of peristaltic pump technology and are designed for simple integration into system control panels for fluid transfer applications.