Masterflex® L/S® Pump Power Supplies, Avantor®
Supplier: Avantor Fluid Handling
MFLA07525-90EA 0 SGD
MFLA07525-90
Masterflex® L/S® Pump Power Supplies, Avantor®
Power Supplies
Optimize performance of your Masterflex pump.
  • Suitable for power requirements 90 to 260 VAC
This replacement power supply is suitable for use with the Masterflex C/L Digital pump.