Masterflex® L/S® Analog Modular Drive Replacement Controllers, Avantor®
Supplier: Avantor Fluid Handling
MFLA07559-04EA 1390 SGD
MFLA07559-04
Masterflex® L/S® Analog Modular Drive Replacement Controllers, Avantor®
Pumps Peristaltic Pumps
Optimize performance of your Masterflex modular drives.
  • Choose replacement controller based on your Masterflex modular system
Extend the life of your Masterflex analog variable-speed modular systems with these replacement parts.