PendoTECH® PressureMAT® Single-Use DPG Handheld Monitors
Supplier: Avantor Fluid Handling
MFLA19406-60EA 0 SGD
MFLA19406-60
PendoTECH® PressureMAT® Single-Use DPG Handheld Monitors
Fluid Sensors
Convenient handheld design allows you to read PendoTECH single-use pressure sensors.
  • Reads sensors to their specification of 0-6 psi +/- 2% of reading; 0-30 psi +/- 3% of reading
  • Battery operated - Uses 9V battery (approximate 14 hours of power)
  • No calibration required when used with the pressure sensors (the standardized sensor output signal is read)
  • One-touch button for sensor tare at 0 psi
  • Minimum / maximum pressure display buttons
  • Auto shut-off after 10 minutes
  • Display to x.x psi with updates >2X per second (x.xx upon request)
These pressureMore Product Information