Pressure Sensor Transmitters
Supplier: Avantor Fluid Handling
MFLA19406-67EA 0 SGD
MFLA19406-67
Pressure Sensor Transmitters
Electrodes Other Electrodes
The enhanced PendoTECH® pressure sensor transmitter connects seamlessly to the PendoTECH pressure sensor, delivering a 4 to 20 mA signal linear with pressure. It maintains its convenient DIN rail mounting design for easy installation and integration into bioprocess systems.
  • The PTR features an alarm output function with a dry contact relay output
  • The PTR supports the digital communication protocol of modbus TCP
  • It has a convenient DIN rail mounting design for easy integration
Two model options are available to deliver optimal performance based onMore Product Information