Flow Control Clamps - Pure-Fit Tru Valve Hosecock
Supplier: Avantor Fluid Handling
MFLA20002-00EA 69 SGD
MFLA20002-00
Flow Control Clamps - Pure-Fit Tru Valve Hosecock
Clamps Tubing and Sanitary Clamps
Easy to implement with installed assemblies and systems!
  • Reusable for multiple cycles and applications
With the Pure-Fit Tru Valve, you no longer need to worry about a time-consuming retrofit to your system. The valve's butterfly design allows easy installation to existing lines; purity is maintained, because the valve goes around the exterior of the tubing and never comes in contact with process fluid. Featuring a simple threaded handle and calibrated scale, the valve ensures accurate control of flow and can also be writtne into validation protocols.More Product Information