Masterflex® Luer Fittings, High-Pressure Stopcock, Avantor®
Supplier: Avantor Fluid Handling
MFLA30526-34EA 128 SGD
MFLA30526-34
Masterflex® Luer Fittings, High-Pressure Stopcock, Avantor®
Valves Ball Valves
Pressure rating up to 1050 psi.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and
More Product Information