Masterflex® Sanitary Tri-Clamp® Screen Gaskets, EPDM, Avantor®
Supplier: Avantor Fluid Handling
30537-04 30537-08 30537-20 30537-22 30537-24 30537-28 30537-40 30537-42 30537-44 30537-48 30537-60 30537-68
MFLA30537-06EA 0 SGD
MFLA30537-06 MFLA30537-26 MFLA30537-46 MFLA30537-62 MFLA30537-64 MFLA30537-66 MFLA30537-80 MFLA30537-82 MFLA30537-84 MFLA30537-86 MFLA30537-88 MFLB30537-04 MFLB30537-08 MFLB30537-20 MFLB30537-22 MFLB30537-24 MFLB30537-28 MFLB30537-40 MFLB30537-42 MFLB30537-44 MFLB30537-48 MFLB30537-60 MFLB30537-68
Masterflex® Sanitary Tri-Clamp® Screen Gaskets, EPDM, Avantor®
Seals Gaskets
Protect your pump, valves and other components in fluid line.
  • Pre-filter before critical membrane filters
  • Generate turbulence for enhanced mass or heat transfer
  • Purify in chromatography applications
Screen gaskets are designed to be interchangeable with standard sanitary clamp gaskets. Gaskets are available in platinum-cured silicone, Viton®, EPDM, PTFE, and Tuf-Steel in mesh sizes of 10, 20, 40, 60 and 100. Screen mesh is constructed of 316L SS. Gaskets meet USP Class VI, USDA, 3-A and cGMP criteria. FDA compliant. All gaskets are animal-derivedMore Product Information