Masterflex® Smart Gasket® 1-Port Sanitary Gaskets, Silicone, Avantor®
Supplier: Avantor Fluid Handling
MFLA30542-20EA 0 SGD
MFLA30542-20
Masterflex® Smart Gasket® 1-Port Sanitary Gaskets, Silicone, Avantor®
Seals Gaskets
Easily sample fluid in critical high-purity system.
  • One internal port accepts thermocouple probe
Masterflex® Smart Gasket® is ideal when validating sterility in a high-purity pharmaceutical system. The Smart Gasket is used to obtain the critical thermal mapping information you need during the validation process. It easily installs between two sanitary standard ferrules.