Masterflex® Smart Gasket® Sanitary Clamps, Stainless Steel, Avantor®
Supplier: Avantor Fluid Handling
30545-19
MFLA30542-19EA 0 SGD
MFLA30542-19 MFLA30543-19 MFLA30544-19 MFLB30545-19 MFLA41001-49 MFLA41001-54
Masterflex® Smart Gasket® Sanitary Clamps, Stainless Steel, Avantor®
Seals Gaskets
Create a secure connection with your Masterflex® Smart Gasket® sanitary gaskets.
  • Constructed of durable stainless steel (SS)
The Smart Gasket clamps are required with the gaskets because they have slots in them that allow for the different number of poarts. The clamps are specific to the number of ports on the gasket.