Smiths Medical® Luer Stopcock Fittings, Polycarbonate
Supplier: Avantor Fluid Handling
MFLA30600-03BLKEA 0 SGD
MFLA30600-03BLK MFLA30600-04BLK MFLA30600-02BLK MFLA30600-01BLK
Smiths Medical® Luer Stopcock Fittings, Polycarbonate
Valves Ball Valves
Regulate flow without any leaking.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery