Masterflex® Adapter Fittings, Female Luer to Threaded, Straight, Avantor®
Supplier: Avantor Fluid Handling
MFLA30800-00EA 97.7 SGD
MFLA30800-00 MFLA45501-60 MFLA45501-66 MFLA45508-60 MFLA45508-66 MFLA45508-76
Masterflex® Adapter Fittings, Female Luer to Threaded, Straight, Avantor®
Fittings and Connectors
Easily transition between luer fittings and UNF or NPT threads.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options
More Product Information