Value Plastics® Adapter Fittings, Female Luer to Hose Barb, Straight, Polypropylene
Supplier: Avantor Fluid Handling
MFLA30800-08BLKEA 860 SGD
MFLA30800-08BLK MFLA45508-00BLK MFLA30800-06BLK
Value Plastics® Adapter Fittings, Female Luer to Hose Barb, Straight, Polypropylene
Fittings and Connectors
Easily transition to a luer fitting.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery