Masterflex® Adapter Fittings, Male Luer to Hose Barb, Straight, Polypropylene, Avantor®
Supplier: Avantor Fluid Handling
MFLA30800-16EA 40.6 SGD
MFLA30800-16 MFLA30800-18 MFLA30800-20 MFLA30800-22 MFLA30800-24 MFLA30800-26 MFLA30800-28 MFLA45518-00 MFLA45518-02 MFLA45518-06 MFLA45518-08 MFLA45518-11 MFLA45518-22 MFLA45518-24 MFLA45518-26 MFLA45518-44
Masterflex® Adapter Fittings, Male Luer to Hose Barb, Straight, Polypropylene, Avantor®
Fittings and Connectors
Easily transition to a luer fitting.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs,
More Product Information