John Guest® Push-to-Connect Check Valves, Acetal
Supplier: Avantor Fluid Handling
MFLA34006-64EA 321 SGD
MFLA34006-64
John Guest® Push-to-Connect Check Valves, Acetal
Fittings and Connectors
Get a secure connections with quick disconnection.
  • Constructed of durable acetal
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing. To connect flexibleMore Product Information