Masterflex® Barbed Panel-Mount High Flow+ Male Quick-Disconnect Fittings, Avantor®
Supplier: Avantor Fluid Handling
MFLA40255-14EA 0 SGD
MFLA40255-14 MFLA40255-44
Masterflex® Barbed Panel-Mount High Flow+ Male Quick-Disconnect Fittings, Avantor®
Fittings and Connectors
Mechanical innovations reduce risk of fluid cross-over and contamination. A mating connection is made with a High+ Flow Female fitting.
  • Aseptic connect and disconnect
  • Spring-less fluid path
  • Recessed mating features
  • USP Class VI Polypropylene and PVDF housing materials
  • Elimination of accidental disconnect with safety slide release
Masterflex® quick-disconnect fittings are an innovative and aseptic connection that allows a larger volume of fluid to pass through at lower line pressures in a smaller body. As the overall quick disconnect design has matured,More Product Information