Masterflex® Single-Use Bag Port 1" Bottle Cap Thread, Avantor®
Supplier: Avantor Fluid Handling
MFLA40331-05EA 0 SGD
MFLA40331-05
Masterflex® Single-Use Bag Port 1" Bottle Cap Thread, Avantor®
Fittings and Connectors
Large opening makes it suitable for high viscosity liquid and powder applications.
  • 38-400 1" Bottle Cap
  • 2-1/4" disc diameter
Masterflex® Single-Use Bag Ports are engineered to be thermal welded or ultrasonic welded to medical and bioreactor bags of similar material. These materials include polyethylene. These bags can be used to transfer fluid, drug compounds, and cell culture in the biopharmaceudical markets. If material validation is needed, please reach out to our techincal support representatives.