Masterflex® Direct-Reading Variable-Area Flowmeters for Argon, Acrylic, Avantor®
Supplier: Avantor Fluid Handling
68561-18 68561-25 68561-32
MFLA68561-14EA 0 SGD
MFLA68561-14 MFLA68561-15 MFLA68561-17 MFLA68561-19 MFLA68561-20 MFLA68561-21 MFLA68561-22 MFLA68561-24 MFLA68561-26 MFLA68561-27 MFLA68561-28 MFLA68561-29 MFLA68561-30 MFLA68561-31 MFLA68561-33 MFLA68561-34 MFLA68561-35 MFLA68561-36 MFLA68561-37 MFLA68561-38 MFLA68561-39 MFLA68561-40 MFLA68561-41 MFLB68561-18 MFLB68561-25 MFLB68561-32
Masterflex® Direct-Reading Variable-Area Flowmeters for Argon, Acrylic, Avantor®
Flow Meters
Designed for labs and plant processes where multiple gases are used.
  • Easy to read English or metric scales
  • Interchangeable scales for routine gases
  • Customer convertible panel to partial or full in-line mounted configurations
  • Panel mounting by means of hex nuts
  • Easy disassembly and assembly for cleaning
Versatile acrylic flowmeters are designed specifically for argon. Flowmeters are offered with or without built-in low hysteresis needle valves. Panel mount flowmeters without valves or use in partial or full in-line mounted configurations. FlowmetersMore Product Information