Masterflex® Open-Head Sensor Retrofit System, Avantor®
Supplier: Avantor Fluid Handling
MFLA77097-00EA 0 SGD
MFLA77097-00
Masterflex® Open-Head Sensor Retrofit System, Avantor®
Pumps Peristaltic Pumps
Instantly shuts down the drive if the pump head is opened during operation.
  • Use in conjunction with Masterflex® L/S® and I/P® open-head sensor equipped pump heads
  • Compatible with Masterflex® L/S® and I/P® pumps without integrated open-head sensor
  • Controller accepts input signals from up to two pump heads
  • Protect technicians and operators tasked with tubing changes and routine maintenance in busy production environments
  • Controller is NEMA 4X / IP66 rated for washdown
The retrofit system is compatible with all I/P® pumps and with L/S® pumps that acceptMore Product Information