Masterflex® B/T® Drive Controller Mounting Bracket, Avantor®
Supplier: Avantor Fluid Handling
MFLA77111-90EA 697 SGD
MFLA77111-90
Masterflex® B/T® Drive Controller Mounting Bracket, Avantor®
Mounting Plates
Make controlling your B/T® drive easier than ever.
  • Suitable for use with B/T modular digital drives
This controller mounting bracket allows a Masterflex® B/T® digital controller to be mounted directly to the pump head for ease of use.