Masterflex® C/L® Pump Power Supply, Avantor®
Supplier: Avantor Fluid Handling
MFLA77120-11EA 0 SGD
MFLA77120-11
Masterflex® C/L® Pump Power Supply, Avantor®
Power Supplies
Safely adapt your VDC powered C/L® pump.
  • Suitable for use with C/L® pumps
Easily convert your 12 VDC C/L® Variable-Speed pumps to 100 to 240 VAC power. This power supply is suitable for use with 12 VDC models 77122-06, -16, and 26 single-channel pumps and models 77120-52 and -62 dual-channel pumps.