Masterflex® I/P® Brushless Process Drive Systems, Avantor®
Supplier: Avantor Fluid Handling
MFLA77965-00EA 0 SGD
MFLA77965-00 MFLA77965-00-UK
Masterflex® I/P® Brushless Process Drive Systems, Avantor®
Pumps Peristaltic Pumps
Powerful, low-maintenance drives are versatile and cost-effective.
  • Flow rate 0.2 to 26 LPM with I/P tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Digital interface with LED display shows percent speed with 1% resolution
  • Speed control via three-turn potentiometer for quick, easy adjustments
  • Easily control start/stop and reverse features via sealed keypad
  • Contructed of non-chip painted steel and aluminum IP55-rated housing
  • Brushless, maintenance-free motor offers ±0.25% speed control accuracy
  • Remote control available using analog
More Product Information