Masterflex® L/S® Digital Process Pump Systems, Avantor®
Supplier: Avantor Fluid Handling
MFLA77975-25-UKEA 0 SGD
MFLA77975-25-UK MFLA77975-45-UK
Masterflex® L/S® Digital Process Pump Systems, Avantor®
Pumps Peristaltic Pumps
Transfer, dispense, and meter in lab, process and washdown environments.
  • Digital interface with graphical LCD shows pump performance and operating mode— continuous run, timed dispense, copy dispense, or volume dispense
  • Remote control available using analog communication—voltage, current and contact closure
  • Dispense by Volume in ml (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle)
  • Antidrip function ensures dispensing accuracy
  • Models with open-head sensor stops
More Product Information