Masterflex® Flow-Through Pump Tubing Weights, Avantor®
Supplier: Avantor Fluid Handling
MFLA07570-04EA 300 SGD
MFLA07570-04 MFLA78226-81 MFLA78226-82
Masterflex® Flow-Through Pump Tubing Weights, Avantor®
Weights Sinkers
Keep your samples flowing and tubing from curling.
  • Choose between PTFE or stainless steel construction
This flow-through tubing weight prevents tubing from curling or floating. Ideal for groundwater sampling, this weight fits Masterflex®; L/S® 15 or L/S® 24 Precision tubing.