Masterflex® I/P® High-Performance Precision Pump Tubing, BioPharm Plus Platinum-Cured Silicone, Avantor®
Supplier: Avantor Fluid Handling
MFLA96441-70EA 0 SGD
MFLA96441-70 MFLA96441-88 MFLA96441-89
Masterflex® I/P® High-Performance Precision Pump Tubing, BioPharm Plus Platinum-Cured Silicone, Avantor®
Tubing Peristaltic Pump Tubing
Up to five times longer life and lower spallation provides enhanced consistency over time.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory Approvals to match your unique application
Choose our BioPharm PlusMore Product Information