Masterflex® Double-Open-End Sediment Filter Cartridges
Supplier: Avantor Fluid Handling
01512-08
MFLB01512-08EA 22.6 SGD
MFLB01512-08
Masterflex® Double-Open-End Sediment Filter Cartridges
Filters Filter Cartridges
Polypropylene fiber medium is thermally bonded to a rigid core for added durability.
  • Constructed of polypropylene (PP)
Ideal for removing a broad range of deformable and nondeformable particles. Rigid design is ideal for filtering high-solids, higher-viscosity fluids that require higher differential pressures. Cartridge imparts no taste, odor, or color. Maximum temperature is 79 °C (175 °F).
Note: For optimal performance, change cartridges at 25 psid.