Battery-Powered Digital Flow Rate Monitor and Totalizer 43643
Supplier: Avantor Fluid Handling
05629-22
MFLB05629-22EA 0 SGD
MFLB05629-22
Battery-Powered Digital Flow Rate Monitor and Totalizer 43643
Flow Controllers
Use in areas where AC power is not readily available.
  • Use in areas where wired power is power is not readily available
Independently calibrate flow and totalizer readings. Monitor includes both fixed and resettable totalizers. Features a four-digit flow LCD, and eight-digit totalizer LCD, automatic contrast adjustment, and low-battery indication. Monitor includes unit/multiplier decals for field calibration.
Operates on two internal lithium batteries (included). Use monitor only with the Roto-X™ and Metalex™ flow sensors sold separately. Note: not forMore Product Information