CPC® Plastic Quick-Disconnect Fittings, Compression Inserts
Supplier: Avantor Fluid Handling
06360-21BLK
MFLB06360-21BLKEA 0 SGD
MFLB06360-21BLK
CPC® Plastic Quick-Disconnect Fittings, Compression Inserts
Fittings and Connectors
Fittings connect in a snap.
  • Constructed of durable acetal plastic—ideal for use with corrosive fluids
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body.
A complete coupler requires a fitting body and an insert (order separately).