Mini Submersible Magnetic Drive Pumps
Supplier: Avantor Fluid Handling
07105-05 07142-02 07142-15 07142-23 07142-17 07142-31
MFLB07105-05EA 0 SGD
MFLB07105-05 MFLB07142-02 MFLB07142-15 MFLB07142-23 MFLB07142-17 MFLB07142-31
Mini Submersible Magnetic Drive Pumps
Pumps Centrifugal Pumps
Operate fully submerged or in open air.
  • Liquid-cooled models keep heat-sensitive fluids from warming up
  • Thick (1/8") epoxy coating dissipates heat quickly and provides good chemical resistance
  • UL-Listed
These mini submersible magnetic drive pumps are good for low-viscosity liquids including solvents, dyes, inks, and mild corrosives.
Delivery information: Includes a three-wire cord.