Gems Bent Stem Float Switches, Stainless Steel
Supplier: Avantor Fluid Handling
07187-22
MFLB07187-22EA 0 SGD
MFLB07187-22
Gems Bent Stem Float Switches, Stainless Steel
Level Sensors
Get longer life and more durability with hermetically sealed switches.
  • Features a 90° stem for side mounting where space is a premium
This bent stem level switch accepts 120/240 VAC and operates as a NO switch. Switches include 24" leads.