Cast iron float and thermostatic steam traps (parallel configuration)
Supplier: Avantor Fluid Handling
07600-30
MFLB07600-30EA 0 SGD
MFLB07600-30
Cast iron float and thermostatic steam traps (parallel configuration)
Filters Gas Filters
Keep steam space free of condensate to provide maximum system efficiency.
  • Constructed of durable cast iron
Float and thermostatic steam traps contain a float valve mechanism which modulates to discharge condensate continuously at steam temperature, while non-condensable gases are released by a separate internal balanced pressure thermosatic air vent. Stainless steel internal parts can be serviced without disturbing the piping connections.
Typical applications:
All process equipment, particularly when controlled by modulating temperature control valves
More Product Information